Learn More
AbD Serotec IgG1 antibody | AbD27686 | 0.1mg
Supplier:  AbD Serotec HCA285
Encompass
Anti-Human IgG1 Fc Antibody recognizes human IgG1 and can be used for the detection of therapeutic antibodies in human IgG1 format in samples obtained from cynomolgus macaques.
Catalog No. NC1768026