Learn More
BIOHIPPO Human APOF Microsample
SDP
Supplier:  BIOHIPPO BHE15212077
Encompass
Human APOF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of APOF in Serum plasma and other biological fluids It provides a detection range of 0 94-60 ng/mL with a sensitivity of 0 44 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human APOF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human APOF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human APOF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15421