Learn More
Abcam Human Apolipoprotein E ELISA Kit (APOE), 1 x 96 Tests
Supplier:  Abcam AB1088131X96TESTS
Encompass_Preferred
Human Apolipoprotein E ELISA Kit (APOE) is a Sandwich ELISA used to quantify Human Apolipoprotein E (APOE) with a sensitivity of 4 ng/ml - Colorimetric ELISA kit - 450 nm readout. Works on any standard plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF) - Wide dynamic range – 0.008 - 0.5 µg/ml - Cited in over 30 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1762746