Learn More
Sino Biological Recombinant Human ASIC1 Protein (VLP, Full-length, His & Strep Tag)
Supplier:  Sino Biological 15953H64H20
Encompass_Preferred
A DNA sequence encoding the Human ASIC1-VLP (Full Length) (P78348-2) (Met1-Cys528) was expressed, with a polyhistidine tag at the N-terminus and a Strep tag at the C-terminus.
Catalog No. 50-299-3522