Learn More
Sino Biological Human BCAM Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEK102385
Encompass_Preferred
The Human BCAM ELISA Pair Set is for the quantitative determination of Human BCAM.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human BCAM coated on a 96-well plate. Standards and samples are added to the wells, and any Human BCAM present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human BCAM monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human BCAM present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85276