Learn More
BIORTUS USA INC Human BCAT2 Protein, 100UG
Supplier:  BIORTUS USA INC BP1724013A
Encompass_Preferred
Gene Name: BCAT2. Uniprot: O15382. Construct: 6His-StrepII-TEV-GG-BCAT2(A28-V392 end). Expression Host: E. coli. Purity: 80%.
Catalog No. NC2953344