Learn More
ACROBiosystems Alexa Fluor 647-Labeled Human BCMA / TNFRSF17 Protein, His TagStar Staining
Supplier:  ACROBiosystems BCAHA2H925UG
Encompass_Preferred
Alexa Fluor 647-Labeled Human BCMA, His Tag (BCA-HA2H9) is produced via conjugation of AF647 to Human BCMA, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human BCMA, His Tag is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).
Catalog No. 50-233-1039
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.