Learn More
ACROBiosystems Recombinant Protein;Human BCMA / TNFRSF17 Protein,His Tag;HEK293;100UG;BCA,TNFRSF17,CD269,BCM,BCMA,TNFRSF-17
Supplier: ACROBiosystems BCAH522Y100UG
Recombinant Protein;Human BCMA / TNFRSF17 Protein,His Tag;100UG;Human BCMA, His Tag (BCA-H522y) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.8 kDa. The protein migrates as 12 kDa and 15-17 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 5 μg/mL (100 μL/well) can bind Human BCMA, His Tag (Cat. No. BCA-H522y) with a linear range of 4-125 ng/mL (QC tested).;Immobilized Human BCMA, His Tag (Cat. No. BCA-H522y) at 2 μg/mL (100 μL/well) can bind Anti-Human BCMA Mab with a linear range of 0.2-1.6 ng/mL (Routinely tested).;Anti-Human BCMA MAb (human IgG1) captured on CM5 chip via Anti-Human IgG Fc antibodies surface, can bind Human BCMA, His Tag (Cat. No. BCA-H522y) with an affinity constant of 32.3 nM as determined in a SPR assay (Biacore T200) (Routinely tested).;293 cells