Learn More
Raybiotech Inc Human BCMA/TNFRSF17 ELISA Kit
Supplier:  Raybiotech Inc ELHTNFRSF171
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 48 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human BCMA.
Catalog No. 50-194-9796