Learn More
American Research Products Inc Human Beta Endorphin b EP ELI
Supplier:  American Research Products Inc CSBE06821H
Encompass
Human Beta-Endorphin b-EP ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum urine cerebrospinal fluid (CSF) saliva tissue homogenates Method Sandwich
Catalog No. 50-003-28817