Learn More
Biomatik Corporation Human Beta-Carotene-15,15'-Monooxygenase 1 (bCMO1) ELISA Kit
Supplier:  Biomatik Corporation EKU02696
Encompass_Preferred
Human Beta-Carotene-15,15'-Monooxygenase 1 (bCMO1) ELISA Kit for detecting bCMO1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.132ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9HAY6. Gene ID: 53630. Target Synonyms: BCO1; BCDO; BCDO1; BCMO; Beta-carotene dioxygenase 1; Beta-carotene oxygenase 1; Beta,beta-carotene 15,15'-monooxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9101