Learn More
ACROBiosystems Human BMP-4 / BMP2B (improved sequence) Protein, premium grade
Supplier:  ACROBiosystems BM4H511820UG
Encompass_Preferred
Bone Morphogenetic Protein 4 (BMP4) is a member of growth factor of the TGF-beta superfamily that plays essential roles in many developmental processes, including neurogenesis, vascular development, angiogenesis and osteogenesis. BMP-4 Initiates the canonical BMP signaling cascade by associating with type I receptor BMPR1A and type II receptor BMPR2. BMP-4 can acts in concert with PTHLH/PTHRP to stimulate ductal outgrowth during embryonic mammary development and to inhibit hair follicle induction.
Catalog No. 50-271-9917