Learn More
Raybiotech Inc C-Series Human BMP Related Array 1 Kit.
Supplier:  Raybiotech Inc AAHBMP18
Encompass_Preferred
Detects 16 Human BMPs. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: BAMBI, BMP-2, BMP-4, BMP-5, BMP-6, BMP-7, BMP-8, BMP-9, BMPR-IA, ALK-6, BMPR-II, COCO, Crossveinless-2, Follistatin, GDF-13
Catalog No. 50-194-8517