Learn More
Raybiotech Inc C-Series BMP Related Array 2 Kit.
Supplier:  Raybiotech Inc AAHBMP28
Encompass_Preferred
Detects 8 Human BMPs. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: GDF-11, BMP-2, BMP-4, BMP-5, BMP-6, BMP-7, BMP-8, BMP-11
Catalog No. 50-194-8520