Learn More
STEMCELL Technologies Human Bone Marrow Stromal Cells Derived in ACF Medium, Frozen, Size: 7.5 x 10^5 cells
SDP
Supplier:  STEMCELL Technologies 70071
Encompass
Human primary stromal cells (mesenchymal stem and progenitor cells) are produced by expanding bone marrow mononuclear cells (MNCs) in culture and cryopreserved following the first passage in culture. Human Bone Marrow Stromal Cells (Catalog #70071) were cultured in MesenCult™-ACF Plus Culture Kit (Catalog #05448).Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.
Catalog No. 50-197-5830