Learn More
Raybiotech Inc Quantibody Human Bone Metabolism Array 2 Kit.
Supplier:  Raybiotech Inc QAHBMA21
Encompass_Preferred
Detects 10 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: BMP-2, BMP-6, BMP-7, DKK-1, MMP-3, Osteoprotegerin (TNFRSF11B), Osteopontin (SPP1), PDGF-BB, TGF beta 3, TRANCE (TNFSF11)
Catalog No. 50-195-0801