Learn More
Amsbio LLC Human Breast Carcinoma and normal adjacent tissue FFPE block
Supplier:  Amsbio LLC TPH1137B
Encompass
Human FFPE tissue blocks are processed from diseased tissues or clinically-unremarkable donor normal human tissue samples. Tissue is prepared by a certified medical pathologist, fixed in 10% neutral formalin for 24 hours and embedded in IHC-grade paraffin. The target FFPE specimen size is 0.5 x 0.5 x 0.5 cm, but it can vary significantly depending on the organ.
Catalog No. 50-204-1347