Learn More
Sino Biological Human C-Reactive Protein Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEKA1125015
Encompass_Preferred
The Human C-Reactive Protein / CRP ELISA Pair Set is for the quantitative determination of Human C-Reactive Protein / CRP.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human C-Reactive Protein / CRP coated on a 96-well plate. Standards and samples are added to the wells, and any Human C-Reactive Protein / CRP present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human C-Reactive Protein / CRP monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human C-Reactive Protein / CRP present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85394