Learn More
ACROBiosystems Human Cadherin-17, His Tag (CA7-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Met 787 (Accession # Q12864-1).
Supplier:  ACROBiosystems CA7H52H31MG
Encompass_Preferred
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 86.9 kDa.
Catalog No. 50-223-1540