Learn More
Sino Biological Recombinant Human CADM1 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11168H08HB100
Encompass_Preferred
A DNA sequence encoding the Human CADM1 (NP_055148.3) (Met1-His374) was expressed, fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-299-3421