Learn More
American Research Products Inc Human calcitonin gene related
Supplier:  American Research Products Inc CSBE08210H
Encompass
Human calcitonin gene related peptide CGRP ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26854