Learn More
Bon Opus Biosciences Recombinant Human Calnexin Protein (C-His) - Source: Human 293 Cells
SDP
Supplier:  Bon Opus Biosciences BP27550UG
Encompass_Preferred
Recombinant Human Calnexin is produced by our Mammalian expression system and the target gene encoding His21-Pro481 is expressed with a 6His tag at the C-terminus Accession#P27824. Formulation: Supplied as a 0.2 um filtered solution of 20mM Tris-HCl, 150mM NaCl, 2mM CaCl2, 10% Glycerol, pH7.5
Catalog No. NC2733759