Learn More
ACROBiosystems ACROBiosystems Human Carbonic Anhydrase IX / CA9 (38-414) Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems CA9H525350UG
Encompass_Preferred
Recombinant Protein;50UG;Human Carbonic Anhydrase IX / CA9 (38-414), Fc Tag (CA9-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession # Q16790-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.4 kDa. The protein migrates as 70-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Carbonic Anhydrase IX / CA9 (38-414), Fc Tag (Cat. No. CA9-H5253) at 1 μg/mL (100 μL/well) can bind Anti-Carbonic Anhydrase IX Antibody, Human IgG1 with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-210-6477