Learn More
Sino Biological Human Carboxypeptidase B1 Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEK105015
Encompass_Preferred
The Human Carboxypeptidase B1 / CPB1 ELISA Pair Set is for the quantitative determination of Human Carboxypeptidase B1 / CPB1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Carboxypeptidase B1 / CPB1 coated on a 96-well plate. Standards and samples are added to the wells, and any Carboxypeptidase B1 / CPB1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Carboxypeptidase B1 / CPB1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Carboxypeptidase B1 / CPB1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are
Catalog No. 50-007-85116