Learn More
Biomatik Corporation Human CAT (Catalase) ELISA Kit
Supplier:  Biomatik Corporation EKF58416
Encompass_Preferred
Human CAT (Catalase) ELISA Kit for detecting CAT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04040. Target Synonyms: CAT; Cas1; Cs-1; catalase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9183