Learn More
BIOHIPPO InVivoMAb Anti-Human CCL14/NCC-2 (Iv0082), 100 µg
Supplier: BIOHIPPO BHA21409914
CCL14/NCC-2 Recombinant Antibody (clone Iv0082) is a recombinant Human IgG1, kappa antibody specific for CCL14/NCC-2. It reacts with Human. Suitable for ELISA, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.