Learn More
Sino Biological Human CCL5/RANTES Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK1090015
Encompass_Preferred
The Human CCL5 / RANTES ELISA Pair Set is for the quantitative determination of Human CCL5 / RANTES.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CCL5 / RANTES coated on a 96-well plate. Standards and samples are added to the wells, and any Human CCL5 / RANTES present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human CCL5 / RANTES monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CCL5 / RANTES present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86643