Learn More
Sino Biological Recombinant Human CCR5 Protein (VLP, Flag & His-Strep Tag)
Supplier:  Sino Biological 13022H82H100
Encompass_Preferred
A DNA sequence encoding the Human CCR5 (P51681) (Met1-Arg319,thermostabilizing mutations) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus.
Catalog No. 50-299-3619