Learn More
Sino Biological Recombinant Human CD10/Neprilysin Protein (His Tag)
Supplier:  Sino Biological 10805H07H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Tyr 52-Trp 750) of human CD10 (NP_000893.2) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-7296