Learn More
ACROBiosystems ACROBiosystems Human CD3 epsilon&CD3 gamma Heterodimer Protein, Fc Tag&Fc Tag (MALS verified)
Supplier:  ACROBiosystems CDGH5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human CD3E&CD3G Heterodimer Protein, Fc Tag&Fc Tag (CDG-H5253) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # NP_000724.1 (CD3E) & AAI13831 (CD3G)).;Human CD3E&CD3G Heterodimer Protein, Fc Tag&Fc Tag is produced by co-expression of CD3E and CD3G, has a calculated MW of 38.3 kDa (CD3E) and 36.9 kDa (CD3G). Subunit CD3E is fused with a human IgG1 Fc tag at the C-terminus and subunit CD3G is fused with a human IgG1 Fc tag at the C-terminus. As a result of glycosylation, the protein migrates as 43-55 kDa under reducing (R) condition, and 90-110 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human CD3E&CD3G Heterodimer Protein, Fc Tag&Fc Tag (Cat. No. CDG-H5253) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3), Ultra-low endotoxin (Cat. No. CDE-M120a) with a linear range of 0.8-3 ng/mL (QC tested).;Immobilized Human CD3E&CD3G Hete
Catalog No. 50-210-6966