Learn More
ACROBiosystems HRP-Human CD3 epsilon&CD3 delta Heterodimer Protein, His Tag&Tag Free
Supplier:  ACROBiosystems CDDHR2W325UG
Encompass_Preferred
HRP-Human CD3E & CD3D Heterodimer Protein, His Tag&Tag Free (CDD-HR2W3) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) & P04234-1 (CD3D)).
Catalog No. 50-233-1851