Learn More
Sino Biological Human CD7 Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEK110285
Encompass_Preferred
The human CD7 ELISA Pair Set is for the quantitative determinationof human CD7.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD7 coated on a 96-well plate. Standards and samples are added to the wells, and any CD7 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-CD7 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of CD7 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85976