Learn More
Biolegend LEGENDplex Human CD8/NK Panel 13-plex, 100 tests, Multiplex bead-based immunoassay, Vial Barcode Plate (VbP), Immune response profiling
Supplier:  Biolegend 741187
Encompass_Preferred
The BioLegend LEGENDplex™ Human CD8/NK Panel (13-plex) is a specialized immunoassay kit designed for the simultaneous quantification of 13 critical human proteins involved in the biology of CD8 T cells and NK cells. This multiplex bead-based assay efficiently conserves sample volume and enhances experimental throughput, making it an excellent solution for immune profiling research. With this kit, researchers can achieve detailed insights into cellular activation, effector functions, and regulatory pathways from a single sample.
  • Multiplex bead-based immunoassay
  • Human reactivity
  • Measures 13 analytes including IL-17A, IL-2, IL-4, IL-10, IL-6, TNF-alpha, Fas, FasL, IFN-gamma, Granulysin, Granzyme A, Granzyme B, and Perforin
  • Available quantity for use: 100 tests
  • Utilizes Vial Barcode Plate (VbP) format
  • Application in immune response profiling
  • Targets CD8 T cells and NK cells
Catalog No. NC2740353