Learn More
Sino Biological Recombinant Human CD81 Protein (VLP, Full-length, Flag Tag)
Supplier:  Sino Biological 14244H25H100
Encompass_Preferred
A DNA sequence encoding the Human CD81-VLP (Full Length) (P60033) (Met1-Tyr236) was fused with a signal peptide at the N-terminus and a flag tag at the C-terminus.
Catalog No. 50-299-2991