Learn More
Sino Biological Human CD83 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK1176315
Encompass_Preferred
The Human CD83 ELISA Pair Set is for the quantitative determination of Human CD83.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CD83 coated on a 96-well plate. Standards and samples are added to the wells, and any Human CD83 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human CD83 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CD83 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86345