Learn More
Cusabio Technology LLC HUMAN CD86 molecule ELISA KIT
Supplier:  Cusabio Technology LLC CSBE08542H
Encompass_Preferred
This kit detects CD86 in human serum plasma and tissue homogenates using a quantitative sandwich ELISA methodology Designed for precise measurement it offers a detection range of 0 312 ng/mL to 20 ng/mL with a sensitivity threshold of 0 078 ng/mL ensuring reliable analysis of low-abundance samples The assay requires a sample volume of 50-100 L and involves an incubation period of 1-5 hours optimized for efficiency without compromising accuracy Absorbance is measured at 450 nm using a standard microplate reader facilitating straightforward integration into laboratory workflows Suitable for research applications requiring specific and reproducible quantification of soluble CD86 this kit provides standardized components and protocols to ensure consistent performance across experimental replicates
Catalog No. 50-000-48475