Learn More
Sino Biological Human CD93 ELISA Pair Set 5PP (Sino Biological)
Supplier:  Sino Biological SEK125895
Encompass_Preferred
Each vial contains 70(KW10AP1104-1) ng of recombinant Human CD93/C1qR. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2000 pg/mL is recommended.
Catalog No. 50-237-0704