Learn More
BIORTUS USA INC Human CDK13/CyclinT1 Protein, 100UG
Supplier:  BIORTUS USA INC BP1646T24A
Encompass_Preferred
Gene Name: CDK13/CCNT1. Uniprot: Q14004/O60563. Construct: GST-TEV-CDK13(Q673-S1054)/His-Flag-TEV-CyclinT1(M1-R259) . Expression Host: Insect cell. Purity: 85%. 1 each contains 100UG.
Catalog No. NC2953298