Learn More
BIORTUS USA INC Human CDK4 Protein, 100UG
Supplier:  BIORTUS USA INC BP2583901A
Encompass_Preferred
Gene Name: CDK4. Uniprot: P11802. Construct:, His-TEV-CDK4(M1-E303 end). Expression Host: Insect cell. Purity: 90%.
Catalog No. NC2953328