Learn More
Abcam Human Cdk6 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Supplier:  Abcam AB25374710X96TESTS
Encompass_Preferred
Human Cdk6 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Cdk6.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-7022