Learn More
Raybiotech Inc G-Series Human Chemokine Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAHCHEG18
Encompass_Preferred
Detects 38 Human Chemokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – AAH-CHE-G1-8
Catalog No. 50-194-8528