Learn More
Sino Biological Recombinant Human Chitotriosidase/CHIT1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11223H08H50
Encompass_Preferred
A DNA sequence encoding the human CHIT1 (NP_003456.1) (Met 1-Asn 466) precursor was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6989