Learn More
Creative Biomart Human chorionic gonadotropin (HCG)
Supplier:  Creative Biomart THP0140
Encompass_Preferred
Human chorionic gonadotropin (HCG), a polypeptide hormone produced by the human placenta. HCG is composed of an alpha and a beta sub-unit. The alpha sub-unit is essentially identical to the alpha sub units of the human pituitary gonadotropins, luteinizing hormone (LH) and follicle-stimulating hormone (FSH), as well as to the alpha sub-unit of human thyroid-stimulating hormone (TSH), while the beta sub units of these hormones differ in amino acid sequence.
Catalog No. 50-208-4191