Learn More
ACROBiosystems Human Claudin-6 / CLDN6 Protein, His,Twin-Strep Tag (Detergent)
Supplier:  ACROBiosystems CL6H528920UG
Encompass_Preferred
Human Claudin-6 Protein, His,Twin-Strep Tag (CL6-H5289) is expressed from Baculovirus-Insect cells. It contains AA Ala 2 - Val 220 (Accession # P56747-1).
Catalog No. 50-233-1625