Learn More
BIOHIPPO Human CLEC11A Microsample
SDP
Supplier:  BIOHIPPO BHE15211109
Encompass
Human CLEC11A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CLEC11A in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 94-60 ng/mL with a sensitivity of 0 41 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CLEC11A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CLEC11A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CLEC11A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13810