Learn More
American Research Products Inc Human Coagulation factor XIII
Supplier:  American Research Products Inc CSBEL007922HU
Encompass
Human Coagulation factor XIII B chain (F13B) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma urine tissue homogenates Method Sandwich
Catalog No. 50-003-27256