Learn More
American Research Products Inc Human Coenzyme Q10 CoQ10 ELI
Supplier:  American Research Products Inc CSBE14081H
Encompass
Human Coenzyme Q10 (CoQ10) ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-50 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26840