Learn More
Chondrex Inc ELISA Grade Human Type I Collagen, 0.5 mg/ml x 1ml
Supplier:  Chondrex Inc 1005
Encompass_Preferred
Highly purified (99% pure by SDS-PAGE) human type I collagen derived from human placenta in 1 mL of 0.05M acetic acid at a concetration of 0.5 mg/ml. Each vial contains enough collagen to coat ten 96-well ELISA plates. It is ideal for use as an ELISA antigen to detect anti-human type I collagen antibodies in serum of experimental animals used in the Collagen-Induced Arthritis (CIA) model, or human serum from patients with arthritis or other diseases. Each vial of ELISA grade human type I collagen also comes with 10 mL of 10X Collagen Dilution Buffer that should be used to prevent the aggregation of collagen molecules when coating ELISA plates. Contact Chondrex, Inc. at support@chondrex.com for technical support.
Catalog No. 50-152-6602