Learn More
Abcam Human Collagen Type I ELISA Kit, 96TEST
Supplier:  Abcam AB28525096TEST
Encompass_Preferred
Human Collagen Type I ELISA kit (colorimetric) is a sandwich ELISA designed to quantify human collagen type I (COL1A1) with a sensitivity of 0.188 ng/mL. - Wide dynamic range – quantifies 0.313–20 ng/mL - Compatible with multiple sample types – works with serum, plasma, tissue homogenates, cell culture supernatants, cell lysates, and other biological fluids - Assay platform – microplate (12 x 8 well strips) for easy setup and reliable results
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5449