Learn More
Biomatik Corporation Human Complexin 1 (CPLX1) ELISA Kit
Supplier:  Biomatik Corporation EKU10746
Encompass_Preferred
Human Complexin 1 (CPLX1) ELISA Kit for detecting CPLX1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O14810. Gene ID: 10815. Target Synonyms: CPX-I; CPX1; Synaphin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6933